Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
far-7
Entrez Gene ID
173421
Gene Name
protein-coding
Gene Aliases
CELE_K01A2.2
Chromosome Location
Chr.II: 319785 - 321445 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061307.7 NP_493708.1 2 - 3 144 86
NR_132574.1 2 - 3 107 86

View Full Product Details