Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-34
Entrez Gene ID
189298
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_W08F4.2
Chromosome Location
Chr.II: 587833 - 589143 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061363.3 NP_493764.2 1 - 2 180 80

View Full Product Details