Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
thoc-1
Entrez Gene ID
173460
Gene Name
THO Complex (transcription factor/nuclear export) subunit
Gene Aliases
CELE_T02H6.2
Chromosome Location
Chr.II: 702107 - 708608 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061395.3 NP_493796.2 1 - 2 82 66

View Full Product Details