Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxc-31
Entrez Gene ID
173472
Gene Name
F-box C protein
Gene Aliases
CELE_R07C3.9
Chromosome Location
Chr.II: 911980 - 913052 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061447.4 NP_493848.1 2 - 3 776 74

View Full Product Details