Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxc-32
Entrez Gene ID
173476
Gene Name
F-box C protein
Gene Aliases
CELE_C32B5.10
Chromosome Location
Chr.II: 946485 - 948191 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061459.4 NP_493860.1 2 - 3 1109 60

View Full Product Details