Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snf-3
Entrez Gene ID
173493
Gene Name
Sodium-and chloride-dependent betaine transporter
Gene Aliases
CELE_T13B5.1
Chromosome Location
Chr.II: 1092006 - 1097905 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061509.9 NP_493910.2 6 - 7 1321 79
GenBank mRNA DQ118731.1 - 6 - 7 1363 79
DQ482733.1 - 6 - 7 1343 79

View Full Product Details