Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- eri-3
- Entrez Gene ID
-
173497
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_W09B6.3
- Chromosome Location
-
Chr.II: 1123534 - 1127791 on Build WBcel235
- Gene Symbol
- taf-6.1
- Entrez Gene ID
-
173498
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_W09B6.2
- Chromosome Location
-
Chr.II: 1125736 - 1132362 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061518.8 |
NP_493919.1 |
5 - 6 |
720 |
111 |
View Full Product Details