Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cogc-4
- Entrez Gene ID
-
173518
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y51H7C.6
- Chromosome Location
-
Chr.II: 1403234 - 1409118 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061565.6 |
NP_493966.1 |
7 - 8 |
2242 |
74 |
- Gene Symbol
- Y51H7C.41
- Entrez Gene ID
-
36805059
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_Y51H7C.41
- Chromosome Location
-
Chr.II: 1402577 - 1409118 on Build WBcel235
View Full Product Details