Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-33
Entrez Gene ID
173533
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_C08E3.3
Chromosome Location
Chr.II: 1604600 - 1606179 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061606.5 NP_494007.1 3 - 4 688 65

View Full Product Details