Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-21
Entrez Gene ID
186647
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_F59H6.8
Chromosome Location
Chr.II: 2014902 - 2026625 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061754.3 NP_494155.1 1 - 2 127 66

View Full Product Details