Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- math-1
- Entrez Gene ID
-
181827
- Gene Name
- MATH domain-containing protein
- Gene Aliases
-
CELE_B0047.4
- Chromosome Location
-
Chr.II: 2046053 - 2046656 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061763.5 |
NP_494164.1 |
1 - 2 |
158 |
82 |
- Gene Symbol
- bath-24
- Entrez Gene ID
-
181826
- Gene Name
- BTB and MATH domain containing
- Gene Aliases
-
CELE_B0047.3
- Chromosome Location
-
Chr.II: 2044619 - 2046750 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061762.3 |
NP_494163.1 |
1 - 2 |
252 |
82 |
View Full Product Details