Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cct-3
Entrez Gene ID
173581
Gene Name
T-complex protein 1 subunit gamma
Gene Aliases
CELE_F54A3.3
Chromosome Location
Chr.II: 2233540 - 2243539 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061817.6 NP_494218.2 2 - 3 217 84

View Full Product Details