Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-71
Entrez Gene ID
191928
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_D2062.3
Chromosome Location
Chr.II: 2627749 - 2629196 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377725.1 NP_001364738.1 3 - 4 505 68

View Full Product Details