Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-11
Entrez Gene ID
185242
Gene Name
SET (trithorax/polycomb) domain containing
Gene Aliases
CELE_F34D6.4
Chromosome Location
Chr.II: 2690472 - 2692576 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377732.1 NP_001364763.1 1 - 2 55 160

View Full Product Details