Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-61
Entrez Gene ID
24104531
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_F34D6.6
Chromosome Location
Chr.II: 2698865 - 2700329 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306582.1 NP_001293511.1 2 - 3 277 155

View Full Product Details