Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srg-21
Entrez Gene ID
189538
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_Y25C1A.9
Chromosome Location
Chr.II: 3088474 - 3091296 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062041.2 NP_494442.2 1 - 2 225 139

View Full Product Details