Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-11
Entrez Gene ID
173719
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_F12E12.7
Chromosome Location
Chr.II: 3751261 - 3752284 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062239.3 NP_494640.2 1 - 2 79 118

View Full Product Details