Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-10
Entrez Gene ID
183607
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_C49D10.4
Chromosome Location
Chr.II: 3869901 - 3873063 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062277.4 NP_494678.3 1 - 2 99 68

View Full Product Details