Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rhgf-2
Entrez Gene ID
173748
Gene Name
protein-coding
Gene Aliases
CELE_T08H4.1
Chromosome Location
Chr.II: 4100785 - 4127849 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381382.1 NP_001368191.1 8 - 9 1637 116
NM_001381384.1 NP_001367992.1 8 - 9 1368 116
NM_001383815.1 8 - 9 1371 116
GenBank mRNA JQ763434.1 - 7 - 8 1211 116

View Full Product Details