Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-2
Entrez Gene ID
188452
Gene Name
protein-coding
Gene Aliases
CELE_T12C9.3
Chromosome Location
Chr.II: 4441103 - 4453151 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001364060.1 NP_001350986.1 21 - 22 3101 69
NM_001364061.1 NP_001350985.1 22 - 23 3314 69
NM_001364062.1 NP_001350987.1 21 - 22 3752 69
NM_001364063.2 NP_001350984.1 22 - 23 3965 69

View Full Product Details