Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- vps-18
- Entrez Gene ID
-
173783
- Gene Name
- Vacuolar protein sorting-associated protein 18 homolog
- Gene Aliases
-
CELE_W06B4.3
- Chromosome Location
-
Chr.II: 4461130 - 4467896 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062387.6 |
NP_494788.3 |
6 - 7 |
2436 |
74 |
View Full Product Details