Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hlh-1
Entrez Gene ID
173788
Gene Name
protein-coding
Gene Aliases
CELE_B0304.1
Chromosome Location
Chr.II: 4519063 - 4522526 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026721.3 NP_001021892.1 2 - 3 452 53
NM_001026722.5 NP_001021893.1 2 - 3 456 53

View Full Product Details