Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-40
Entrez Gene ID
188086
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_T05A7.8
Chromosome Location
Chr.II: 4681715 - 4683793 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062429.3 NP_494830.2 5 - 6 967 133

View Full Product Details