Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gst-9
Entrez Gene ID
187615
Gene Name
Probable glutathione S-transferase 9
Gene Aliases
CELE_R05F9.5
Chromosome Location
Chr.II: 4891545 - 4893573 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027095.2 NP_001022266.1 1 - 2 181 116

View Full Product Details