Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sgt-1
Entrez Gene ID
173846
Gene Name
Small Glutamine-rich Tetratrico repeat protein
Gene Aliases
CELE_R05F9.10
Chromosome Location
Chr.II: 4902222 - 4903802 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062492.7 NP_494893.1 2 - 3 848 80

View Full Product Details