Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gst-30
Entrez Gene ID
191347
Gene Name
Glutathione S-Transferase
Gene Aliases
CELE_ZK546.11
Chromosome Location
Chr.II: 4933883 - 4935054 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062501.4 NP_494902.1 1 - 2 172 88

View Full Product Details