Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ulp-2
Entrez Gene ID
173859
Gene Name
protein-coding
Gene Aliases
CELE_Y38A8.3
Chromosome Location
Chr.II: 4955450 - 4959876 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318311.3 NP_001305240.1 2 - 2 153 74
NM_062513.5 NP_494914.1 1 - 2 153 74

View Full Product Details