Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lrr-1
Entrez Gene ID
173869
Gene Name
Leucine-Rich Repeat protein
Gene Aliases
CELE_F33G12.4
Chromosome Location
Chr.II: 5034142 - 5036439 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062527.5 NP_494928.1 7 - 8 1282 89

View Full Product Details