Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gur-5
Entrez Gene ID
191665
Gene Name
GUstatory Receptor family
Gene Aliases
CELE_ZK622.2
Chromosome Location
Chr.II: 5285329 - 5288908 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062591.6 NP_494992.2 5 - 6 566 145

View Full Product Details