Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
agt-2
Entrez Gene ID
184244
Gene Name
AlkylGuanine DNA alkylTransferase
Gene Aliases
CELE_F09E5.13
Chromosome Location
Chr.II: 5371980 - 5373827 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062607.5 NP_495008.1 3 - 4 959 85

View Full Product Details