Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hlh-26
Entrez Gene ID
173939
Gene Name
Helix-loop-helix protein 26
Gene Aliases
CELE_C17C3.8
Chromosome Location
Chr.II: 5532836 - 5534053 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062661.6 NP_495062.1 3 - 4 533 122

View Full Product Details