Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acdh-2
Entrez Gene ID
173940
Gene Name
protein-coding
Gene Aliases
CELE_C17C3.12
Chromosome Location
Chr.II: 5544778 - 5547492 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383838.1 2 - 3 273 126

View Full Product Details