Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zyg-1
Entrez Gene ID
173956
Gene Name
Probable serine/threonine-protein kinase zyg-1
Gene Aliases
CELE_F59E12.2
Chromosome Location
Chr.II: 5649748 - 5654422 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062702.6 NP_495103.1 4 - 5 1343 129
GenBank mRNA AF285179.1 - 4 - 5 1365 129

View Full Product Details