Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
casy-1
Entrez Gene ID
174006
Gene Name
protein-coding
Gene Aliases
CELE_B0034.3
Chromosome Location
Chr.II: 5944851 - 5956683 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025939.6 NP_001021110.1 10 - 11 2701 74
NM_001047257.6 NP_001040722.1 2 - 3 190 74
NM_062788.7 NP_495189.2 3 - 4 391 74
GenBank mRNA AY091591.1 - 10 - 11 2662 74

View Full Product Details