Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
casy-1
Entrez Gene ID
174006
Gene Name
protein-coding
Gene Aliases
CELE_B0034.3
Chromosome Location
Chr.II: 5944851 - 5956683 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025939.6 NP_001021110.1 2 - 3 224 65
GenBank mRNA AY091591.1 - 2 - 3 185 65

View Full Product Details