Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mps-1
Entrez Gene ID
183012
Gene Name
MiRP K channel accessory Subunit
Gene Aliases
CELE_C29F5.4
Chromosome Location
Chr.II: 6292452 - 6294361 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062862.5 NP_495263.2 3 - 4 531 125

Gene Symbol
C29F5.3
Entrez Gene ID
183011
Gene Name
RNase_Zc3h12a domain-containing protein
Gene Aliases
CELE_C29F5.3
Chromosome Location
Chr.II: 6290581 - 6294339 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062861.5 NP_495262.2 3 - 4 509 125

View Full Product Details