Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-75
Entrez Gene ID
186968
Gene Name
Putative cuticle collagen 75
Gene Aliases
CELE_K03H9.2
Chromosome Location
Chr.II: 6426293 - 6427513 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062893.2 NP_495294.1 1 - 2 279 54

View Full Product Details