Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-95
Entrez Gene ID
185633
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_F41G3.11
Chromosome Location
Chr.II: 6753126 - 6754774 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062979.3 NP_495380.2 6 - 7 694 117

View Full Product Details