Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-5
Entrez Gene ID
184302
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_F10E7.10
Chromosome Location
Chr.II: 7113683 - 7115906 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063071.2 NP_495472.1 5 - 6 1011 64

View Full Product Details