Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prx-13
Entrez Gene ID
174192
Gene Name
Probable peroxisomal membrane protein PEX13
Gene Aliases
CELE_F32A5.6
Chromosome Location
Chr.II: 7250151 - 7252015 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063112.4 NP_495513.1 3 - 4 417 128

View Full Product Details