Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aka-1
Entrez Gene ID
174216
Gene Name
protein-coding
Gene Aliases
CELE_D1022.7
Chromosome Location
Chr.II: 7472606 - 7478539 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026874.4 NP_001022045.1 13 - 14 3413 114
NM_001383888.1 14 - 15 3314 114
NM_063164.8 NP_495565.2 13 - 14 3425 114
GenBank mRNA AF045926.1 - 13 - 14 3424 114

View Full Product Details