The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001026874.4 | NP_001022045.1 | 8 - 9 | 2551 | 75 |
NM_001383888.1 | 9 - 10 | 2438 | 75 | ||
NM_063164.8 | NP_495565.2 | 8 - 9 | 2563 | 75 | |
GenBank mRNA | AF045926.1 | - | 8 - 9 | 2562 | 75 |