Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wrt-1
Entrez Gene ID
174223
Gene Name
Warthog protein 1 N-product
Gene Aliases
CELE_ZK1290.12
Chromosome Location
Chr.II: 7530996 - 7533246 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063178.8 NP_495579.3 4 - 5 1360 111

View Full Product Details