Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wrt-10
Entrez Gene ID
174224
Gene Name
WaRThog (hedgehog-like family)
Gene Aliases
CELE_ZK1290.8
Chromosome Location
Chr.II: 7533262 - 7534563 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063179.5 NP_495580.1 2 - 3 539 64

View Full Product Details