Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spdl-1
Entrez Gene ID
174256
Gene Name
Spindly-like protein spdl-1
Gene Aliases
CELE_C06A8.5
Chromosome Location
Chr.II: 7781566 - 7783354 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063236.6 NP_495637.1 2 - 3 301 78

View Full Product Details