Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-17
Entrez Gene ID
174269
Gene Name
Zinc finger transcription factor family protein 17
Gene Aliases
CELE_T09A5.12
Chromosome Location
Chr.II: 7866256 - 7870168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063255.7 NP_495656.2 2 - 3 165 79

View Full Product Details