Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gei-14
Entrez Gene ID
174328
Gene Name
GEX Interacting protein
Gene Aliases
CELE_K01C8.5
Chromosome Location
Chr.II: 8273543 - 8276231 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063344.4 NP_495745.2 3 - 4 1113 104

View Full Product Details