Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ctg-2
Entrez Gene ID
174360
Gene Name
protein-coding
Gene Aliases
CELE_T13H5.2
Chromosome Location
Chr.II: 8515054 - 8516922 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267227.3 NP_001254156.1 3 - 4 599 121
NM_001381528.1 NP_001367210.1 3 - 4 563 121

View Full Product Details