Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-38
Entrez Gene ID
174370
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_F54C9.4
Chromosome Location
Chr.II: 8568035 - 8569206 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063409.6 NP_495810.1 1 - 2 279 146

View Full Product Details