Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpf-1
Entrez Gene ID
174378
Gene Name
WD_REPEATS_REGION domain-containing protein
Gene Aliases
CELE_F28C6.3
Chromosome Location
Chr.II: 8600449 - 8602479 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063421.6 NP_495822.1 3 - 4 581 93

View Full Product Details